Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC)

Leverandør: AAT BIOQUEST
103801C0 103801C1 103801C2
AATB103801C0EA 3175 NOK
AATB103801C0 AATB103801C1 AATB103801C2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC)
Antistoffer
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD38 CD38
Environmentally Preferable
Antigen-navn CD38 antigen CD38 antigen
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering APC (Allophycocyanin) APC (Allophycocyanin)
Klon HIT2 HIT2
Reaktivitet Human
Vert Mouse Mouse
Isotype IgG1 IgG1
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp 2...8 °C with minimised light exposure. Do not freeze. 2...8 °C with minimised light exposure. Do not freeze.
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR