iFluor® 675 succinimidyl ester
Leverandør: AAT BIOQUEST
1043
AATB1043EA 5025 NOK
AATB1043
iFluor® 675 succinimidyl ester
iFluor® 675 succinimidyl ester
iFluor® 675 dyes have fluorescence excitation and emission maxima of ~677 nm and ~710 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
NB: For Research Use Only