iFluor® 700, Amine
Leverandør: AAT BIOQUEST
Warning
1077
AATB1077EA 3600 NOK
AATB1077
iFluor® 700, Amine
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3b031d03ac643fb97c91fa474f4e88101540651&vid=e83a337bd4b5a35cf033ca073b502eb1407c9f06&prid=web&clid=SAPDAM