iFluor® 405, Hydrazide

Leverandør: AAT BIOQUEST
Warning

1081
AATB1081EA 3600 NOK
AATB1081
iFluor® 405, Hydrazide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d49ae9cecd7a9d07870fa15aa3256f1867ab8816&vid=fe4b4f856f9e2a7678c37d2125a06f080c58a03e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR