ReadiUse™ Preactivated HRP Maleimide
Leverandør: AAT BIOQUEST
11026
AATB11026EA 1800 NOK
AATB11026
ReadiUse™ Preactivated HRP Maleimide
Assay Immunologisk analyser
ReadiUse™ Preactivated maleimide is a thiol-reactive HRP derivative.
  • Ready-to-use
  • Highly useful antibody label
  • Widely used for immunoassays
  • Multiple applications
NB: For research use only.