iFluor® 633 tyramide
Leverandør: AAT BIOQUEST
11083
AATB11083EA 3325 NOK
AATB11083
iFluor® 633 tyramide
iFluor® 633 tyramide
iFluor® 633 tyramide is an ideal probe for applications where the common tyramides may face interference due to the inherent fluorescence of tissues or other samples.
  • Stable for 12 months
For many immunohistochemical (IHC) applications, traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
NB: For Research Use Only