Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Leverandør: AAT BIOQUEST
11321
AATB11321EA 4725 NOK
AATB11321
Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Assay Enzymanalyser
Designed for consistent and dependable results. The Amplite® colorimetric GAPDH activity assay kit is a sensitive and simple tool for monitoring GAPDH activity based on a coupled enzymatic reaction catalysed by GAPDH.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.