Amplite® Fluorimetric DPP4 Activity Assay Kit
Leverandør: AAT BIOQUEST
11323
AATB11323EA 7875 NOK
AATB11323
Amplite® Fluorimetric DPP4 Activity Assay Kit
Assay Enzymanalyser
Designed for consistent and dependable results. The Amplite® fluorimetric DPP4 activity assay kit provides a simple, quick, and direct protocol for measuring DPP4 Activity.
  • Ex/Em = 360/460 nm
  • Assay is compatible with high-throughput screening systems
The fluorescence intensity generated is directly proportional to the DPP4 activity in the sample, making it suitable for detecting DPP4 in various biological matrices such as cellular lysates, tissue extracts and serum.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.