Amplite® Fluorimetric DPP4 Inhibitor Screening Kit
Leverandør: AAT BIOQUEST
11324
AATB11324EA 7875 NOK
AATB11324
Amplite® Fluorimetric DPP4 Inhibitor Screening Kit
Assay Enzymanalyser
Designed for consistent and dependable results. The Amplite® Fluorimetric DPP4 Inhibitor Screening Kit provides an efficient and direct method for evaluating potential DPP4 inhibitors.
  • Ex/Em=360/460 nm
  • Assay is optimised for high-throughput screening systems
This kit includes Sitagliptin, a well-characterised DPP4 inhibitor used in diabetes treatment, as a reference control, facilitating the assessment of inhibitor efficacy.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.