Violet 510 acid, mFluor™ fluorescent probe

Leverandør: AAT BIOQUEST
Warning

1141
AATB1141EA 5400 NOK
AATB1141
Violet 510 acid, mFluor™ fluorescent probe
Violet 510 acid

AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 500 kDa
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2dd555144edd774eeff6bb1d40dfd86f193e2f8&vid=13ca9548712b4dc38a29b7810f9aab7a37597ade&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR