Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 350)

Leverandør: AAT BIOQUEST
11610010 11610011
AATB11610010EA 4350 NOK
AATB11610010 AATB11610011
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 350)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 350
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR