Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 790)

Leverandør: AAT BIOQUEST
116100M0 116100M1
AATB116100M0EA 4350 NOK
AATB116100M0 AATB116100M1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 790)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 790
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD161
Environmentally Preferable
Antigen-navn CD161 antigen
Antistofftype Primary
Klonalitet Monoclonal
Konjugering iFluor® 790
Klon HP-3G10
Reaktivitet Human
Vert Mouse
Isotype
ImmunoFluorescens Yes
Flowcytometri Yes
Rensing Affinity chromatography
Kyssabsorpsjon No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR