Amplite® Colorimetric Gama-Glutamyltransferase (GGT) Activity Assay Kit
Leverandør: AAT BIOQUEST
11801
AATB11801EA 7875 NOK
AATB11801
Amplite® Colorimetric Gama-Glutamyltransferase (GGT) Activity Assay Kit
Assay Enzymanalyser
Designed for consistent and dependable results. The Amplite® Colorimetric Gamma-Glutamyltransferase (GGT) Activity Assay Kit provides an efficient and straightforward method for measuring GGT activity across various sample types.
  • Robust protocol
  • Absorbance measured at 418 nm
  • Six-month shelf life
It is compatible with a wide range of biological samples, including cells, tissue extracts, and serum, and is suitable for high-throughput systems.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.