Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Leverandør: AAT BIOQUEST
11802
AATB11802EA 9500 NOK
AATB11802
Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Assay Enzymanalyser
Designed for consistent and dependable results. The Screen Quest™ fluorimetric neutrophil elastase inhibitor screening kit offers a fast, sensitive, and high-throughput method for screening and characterising potential inhibitors of NE.
  • Ex/Em=360/470nm
  • Consistent results
  • Six-month shelf life
The amount of fluorescent product is directly proportional to the enzymatic activity of the neutrophil elastase present in the sample.
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.