Red 780 succinimidyl ester, mFluor™ fluorescent probe

Leverandør: AAT BIOQUEST
Warning

Synonymer: Red 780 SE

1191
AATB1191EA 5400 NOK
AATB1191
Red 780 succinimidyl ester, mFluor™ fluorescent probe
Red 780 succinimidyl ester

mFluor™ Red 780 dyes are an excellent alternative to APC-Alexa Fluor® 750 tandems since they have the spectral properties equivalent to those of APC-Alexa Fluor® 750 conjugates.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 1328.65 g/mol
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR