Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (iFluor® 700)

Leverandør: AAT BIOQUEST
120000J0
AATB120000J0EA 4350 NOK
AATB120000J0
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104] (iFluor® 700)
Antistoffer
OX-104 is an anti-human monoclonal antibody that forms an immune complex with the CD200 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD200 antigen
Clonality: Monoclonal
Clone: OX-104
Conjugation: iFluor® 700
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR