ReadiLink™ Rapid iFluor® Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Leverandør: AAT BIOQUEST
1250
AATB1250EA 2700 NOK
AATB1250
ReadiLink™ Rapid iFluor® Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Systemer for proteinkobling og -merking
AAT Bioquest's iFluor® dyes are developed for labelling proteins, in particular, antibodies.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
NB: For research use only.