ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Leverandør: AAT BIOQUEST
1255
AATB1255EA 2700 NOK
AATB1255
ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Systemer for proteinkobling og -merking
Although FITC is still the most popular fluorescent labelling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
NB: For research use only.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vert