5(6)-FAM cadaverine ≥95%
Leverandør: AAT BIOQUEST
127
AATB127EA 3600 NOK
AATB127
5(6)-FAM cadaverine ≥95%
5(6)-FAM cadaverine
FAM cadaverine is an excellent building block for developing fluoresceinated bioconjugates.
  • Multiple Bioconjugation Applications
  • Excellent Quality
NB: For research use only.