ReadiLink™ Rapid trFluor™ Eu Antibody Labeling Kit "Microscale Optimised for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Leverandør: AAT BIOQUEST
1300
AATB1300EA 17325 NOK
AATB1300
ReadiLink™ Rapid trFluor™ Eu Antibody Labeling Kit "Microscale Optimised for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Systemer for proteinkobling og -merking
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Easy and simple protocol for Eu labeling
  • TRF assay development
  • Large Stokes shift
NB: For research use only.