Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Leverandør: AAT BIOQUEST
1302
AATB1302EA 20375 NOK
AATB1302
Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Systemer for proteinkobling og -merking
Microscale optimised for labelling 100 µg antibody per reaction. Many biological compounds present in cells, serum, or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
Designed to provide high quality labelled antibodies for multiple downstream applications.
Leveringsinformasjon: Shipped at RT.
NB: Research use only.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vert