ApoSight™ Green Caspase 3/7 substrate
Leverandør: AAT BIOQUEST
13290
AATB13290EA 3425 NOK
AATB13290
ApoSight™ Green Caspase 3/7 substrate
Assay Proteinanalyser
ApoSight™ Green Caspase 3/7 substrate is the first fluorogenic probe for the direct fluorescence imaing of caspase activities in live cells.
  • unique fluorogenic caspase activity probe for live cells
  • sensitive and accurate detection
  • can be adapted for automation
NB: Storage: Freeze (< −15 °C)