Ac-DEVD-AMC
Leverandør: AAT BIOQUEST
13402
AATB13402EA 1800 NOK
AATB13402
Ac-DEVD-AMC
Proteiner
Ac-DEVD-AMC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning