Native Casein (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Leverandør: AAT BIOQUEST
13440
AATB13440EA 3600 NOK
AATB13440
Native Casein (from Eukaryotic cells), FITC (Fluorescein Isothiocyanate)
Proteiner
Casein is considered to be a generic substrate for a broad spectrum of proteases.
  • Excellent fluorescence intensity change
  • Signal directly proportional to protease activity
  • Bright signal
NB: For research use only.