iFluor® 790, Acid
Leverandør: AAT BIOQUEST
1360
AATB1360EA 3600 NOK
AATB1360
iFluor® 790, Acid
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright fluorescence
  • High photostability
  • Low quenching on proteins
  • Useful for in-vivo imaging
NB: For research use only.