AMP-Fluorescein conjugate calibrator
Leverandør: AAT BIOQUEST
13610
AATB13610EA 5400 NOK
AATB13610
AMP-Fluorescein conjugate calibrator
AMP-Fluorescein conjugate calibrator
AMP-Fluorescein conjugate calibrator is desigend to use with AAT Bioquests' green fluorescent FAM-cAMP PDE IV substrate, a cAMP derivative that is a specific substrate for phosphodiesterase (PDE) IV.
  • Dependable hydrolysis calibration
  • Steady signal
NB: For research use only.