GMP-Fluorescein conjugate for calibration
Leverandør: AAT BIOQUEST
Warning
13611
AATB13611EA 9000 NOK
AATB13611
GMP-Fluorescein conjugate for calibration
GMP-Fluorescein
GMP-Fluorescein conjugate calibrator is desigend to use with green fluorescent FAM-cGMP PDE V substrate, a cGMP derivative that is a specific substrate for phosphodiesterase (PDE) V.
  • Pde V specific
  • High quality
  • Multiple applications
NB: For research use only.
ADR: 5,II