iFluor® 790, Reactive Form_Maleimide
Leverandør: AAT BIOQUEST
1366
AATB1366EA 5400 NOK
AATB1366
iFluor® 790, Reactive Form_Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • Useful For In-Vivo Imaging
NB: For research use only.