4-Methylumbelliferyl-β-D-glucopyranoside ≥95%

Leverandør: AAT BIOQUEST
Warning

Synonymer: 4-Methylumbelliferyl beta-D-Glucopyranoside, 4-Methylumbelliferyl β-D-Glucopyranoside, 4-MU-β-D-Glc, 4-Methyl-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, MUGlucosid

14014
AATB14014EA 1425 NOK
AATB14014
4-Methylumbelliferyl-β-D-glucopyranoside ≥95%
4-Methylumbelliferyl-beta-D-glucopyranoside

4-Methylumbelliferyl-beta-D-glucoside is a fluorogenic MU substrate for detecting glucosidase in cell extracts and purified enzyme preparations using a fluorescence microplate reader or fluorometer.

Formel: C₁₆H₁₈O₈
MW: 338.312 g/mol
MDL nummer: MFCD00063694
CAS-nummer: 18997-57-4
ADR: 5 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR