iFluor® 860, Maleimide
Leverandør: AAT BIOQUEST
Warning
1408
AATB1408EA 5400 NOK
AATB1408
iFluor® 860, Maleimide
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47d6adedf65abb981373d36f646bb80409cfa509&vid=6f0756181f0bec9ccb5a7271906c4a3fd438df52&prid=web&clid=SAPDAM