ReadiUse™ NADP Regenerating Kit
Leverandør: AAT BIOQUEST
15266
AATB15266EA 4550 NOK
AATB15266
ReadiUse™ NADP Regenerating Kit
Assay Enzymanalyser
Designed for consistent and dependable results. AAT Bioquest's ReadiUse™ NADP regenerating kit provides a flexible prepackaged NADP regenerating system to accommodate various experimental designs.
  • Uses two ready-to-use solutions to regenerate NADP by a simple mixing
  • 300-500 enzyme assays can be performed using this kit
  • Six-month shelf life
This kit can be used for all NADP-requiring systems (such as: 12-Ketochenodeoxycholic Acid synthesis, Ferredoxin Reductase System).
Leveringsinformasjon: Shipped with blue ice.
NB: Research use only.