iFluor® 665, Succinimidyl Ester fluorescent probe

Leverandør: AAT BIOQUEST
Warning

1550 1551 1552 1553
AATB1550EA 1625 NOK
AATB1550 AATB1551 AATB1552 AATB1553
iFluor® 665, Succinimidyl Ester fluorescent probe
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • High photostability
  • Brighter signal

ADR: 5,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR