MTS reagent 5 mM vandig løsning, ReadiUse™

Leverandør: AAT BIOQUEST

15710 15711
AATB15710EA 2925 NOK
AATB15710 AATB15711
MTS reagent 5 mM vandig løsning, ReadiUse™
MTS reagent

MTS reagent is one of the most common water-soluble tetrazolium reagents that are widely used to quantify cell viability by absorption spectrophotometry.


  • Stable for 6 months


Quality tested in-house


NB: For Research Use Only

Formel: C₂₀H₁₇N₅O₆S₂
CAS-nummer: 138169-43-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR