mFluor™ Violet 530 maleimide
Leverandør: AAT BIOQUEST
1617
AATB1617EA 4925 NOK
AATB1617
mFluor™ Violet 530 maleimide
mFluor™ Violet 530 maleimide
mFluor™ Violet 530 dyes have fluorescence excitation and emission maxima of ~405 nm and ~530 nm respectively.
  • Stable for 12 months
Quality tested in-house
NB: For Research Use Only