mFluor™ UV455 SE
Leverandør: AAT BIOQUEST
1642
AATB1642EA 5025 NOK
AATB1642
mFluor™ UV455 SE
mFluor™ UV455 SE
mFluor™ UV 455 dyes have fluorescence excitation and emission maxima of ~350 nm and ~455 nm respectively.
  • Stable for 12 months
mFluor™ dyes are developed for multicolor flow cytometry-focused applications.