Anti-IgG Goat Polyclonal Antibody (trFluor™ Tb)
Leverandør: AAT BIOQUEST
16519
AATB16519EA 5450 NOK
AATB16519
Anti-IgG Goat Polyclonal Antibody (trFluor™ Tb)
Antistoffer
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
This trFluor™ Tb goat anti-mouse IgG (H+L) conjugate is commonly used as a second step reagent for indirectMore Product Information