Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 750)

Leverandør: AAT BIOQUEST
16720 16842
AATB16720EA 1800 NOK
AATB16720 AATB16842
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 750)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

NB: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Antigen-navn Immunoglobulin G
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering iFluor® 750
Klon IgG
Reaktivitet Rabbit
Vert Goat
Isotype IgG (H, L)
Kyssabsorpsjon Min×Human

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR