Anti-IgG Goat Polyclonal Antibody (iFluor® 532)

Leverandør: AAT BIOQUEST
16536 16775
AATB16536EA 1800 NOK
AATB16536 AATB16775
Anti-IgG Goat Polyclonal Antibody (iFluor® 532)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

NB: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor™ 532
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR