Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 350)

Leverandør: AAT BIOQUEST
16600 16795
AATB16600EA 1425 NOK
AATB16600 AATB16795
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 350)
Antistoffer
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

NB: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 350
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR