Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)

Leverandør: AAT BIOQUEST
16856
AATB16856EA 2700 NOK
AATB16856
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Cy7®)
Antistoffer
AAT Bioquest's anti-mouse secondary antibodies are affinity-purified antibodies with well-characterised specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity-purified
  • Bright signal

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Cy7®
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR