Anti-IgG Goat Polyclonal Antibody [clone: IgG] (APC (Allophycocyanin)/Cy7®)

Leverandør: AAT BIOQUEST
16873
AATB16873EA 3600 NOK
AATB16873
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (APC (Allophycocyanin)/Cy7®)
Antistoffer
AAT Bioquest's anti-rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity-purified
  • High specificity and affinity
  • Improved sensitivity
  • Bright signal

NB: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR