DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
Leverandør: AAT BIOQUEST
17034
AATB17034EA 6250 NOK
AATB17034
DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
Nukleinsyre reagenser Nukleotider
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
  • Chemically identical to Diethylaminocoumarin-5-dUTP (Perkin Elmer’s NEL455001EA)
This Coumarin-dUTP conjugate can be used as a blue fluorescence colour with Spectrum Aqua™ filter set (Spectrum Aqua™ is the trademark of Vysis).
NB: For research use only.