Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Leverandør: AAT BIOQUEST
17035
AATB17035EA 1625 NOK
AATB17035
Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Nukleinsyre reagenser Nukleotider
Fluorescein-12-dCTP might be incorporated into a DNA sequence as its natural counterpart dCTP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labelling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and cytidine has been optimised for the optimal substrate incorporation and labelling efficiency.
NB: For research use only.