iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Leverandør: AAT BIOQUEST
17041
AATB17041EA 1625 NOK
AATB17041
iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7,5)*
Nukleinsyre reagenser Nukleotider
This iFluor® 488-PEG12-dUTP conjugate has the same spectral properties to ChromaTide™ Alexa Fluor 488-dUTP.
  • PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
NB: For research use only.