XFD488 amine fluorescent probe

Leverandør: AAT BIOQUEST
Warning

1705
AATB1705EA 5400 NOK
AATB1705
XFD488 amine fluorescent probe
XFD488 amine

XFD488 amine is the same molecule to Alexa Fluor® 488 C5 amine.


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 719,83 g/mol
ADR: 5,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR