MagaDye™ 535 ddGTP
Leverandør: AAT BIOQUEST
17063 17067
AATB17063EA 16500 NOK
AATB17063 AATB17067
MagaDye™ 535 ddGTP
Nukleinsyre reagenser Sekvenseringsreagenser
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
NB: For research use only.