XFD594 amine fluorescent probe

Leverandør: AAT BIOQUEST
Warning

1715
AATB1715EA 5400 NOK
AATB1715
XFD594 amine fluorescent probe
XFD594 amine

XFD594 amine is the same molecule to Alexa Fluor® 594 C5 amine (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

MW: 908,14 g/mol
ADR: 5,II
New Product

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR