ddTTP [2',3'-Dideoxythymidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Leverandør: AAT BIOQUEST
17208
AATB17208EA 3425 NOK
AATB17208
ddTTP [2',3'-Dideoxythymidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Nukleinsyre reagenser Sekvenseringsreagenser
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
NB: For research use only.