ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
Leverandør: AAT BIOQUEST
17300
AATB17300EA 1600 NOK
AATB17300
ReadiView™ Blue Real-Time PCR Visualisation Dye *200X*
Nukleinsyre reagenser Nucleic Acid Modification and Labelling
ReadiView™ Blue Real-Time PCR Visualisation Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
  • Stable for 12 months
Quality tested in-house.
NB: For research use only.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon