Nuclear Orange™ LCS1
Leverandør: AAT BIOQUEST
17541
AATB17541EA 1800 NOK
AATB17541
Nuclear Orange™ LCS1
Assay Cellulær analyse
The Nuclear Orange™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analyzing DNA content in living cells.
  • Cell-Permeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
5 mM DMSO Solution
NB: For research use only.